2,5-Dimethylphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-2669-5G BB-2669-25G BB-2669-100G
COBBBB-2669-5GEA 0 GBP
COBBBB-2669-5G COBBBB-2669-25G COBBBB-2669-100G
2,5-Dimethylphenylboronic acid
2,5-Dimethylphenylboronic acid
Formula: C₈H₁₁BO₂
MW: 149.98 g/mol
MDL Number: MFCD01863525
CAS Number: 85199-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR