2,5-Difluorobenzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

CA-4075-5G CA-4075-25G CA-4075-100G
COBBCA-4075-5GEA 0 GBP
COBBCA-4075-5G COBBCA-4075-25G COBBCA-4075-100G
2,5-Difluorobenzoic acid
2,5-Difluorobenzoic acid
Formula: C₇H₄F₂O₂
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: 131…132 °C
MDL Number: MFCD00002410
CAS Number: 2991-28-8
EINECS: 221-060-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR