2,5-Dibromothiazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6658-5G HC-6658-25G HC-6658-1G
COBBHC-6658-5GEA 0 GBP
COBBHC-6658-5G COBBHC-6658-25G COBBHC-6658-1G
2,5-Dibromothiazole
2,5-Dibromothiazole
Formula: C₃HBr₂NS
MW: 242.92 g/mol
MDL Number: MFCD00016891
CAS Number: 4175-78-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR