2,5-Dibromopyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OT-0325-25G OT-0325-100G
COBBOT-0325-25GEA 0 GBP
COBBOT-0325-25G COBBOT-0325-100G
2,5-Dibromopyridine
2,5-Dibromopyridine
Formula: C₅H₃Br₂N
MW: 236.89 g/mol
Boiling Pt: 234…235 °C (1013 hPa)
Melting Pt: 90…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00006221
CAS Number: 624-28-2
EINECS: 210-839-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR