2,4-Dimethylphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

BB-2786-25G BB-2786-5G BB-2786-1G
COBBBB-2786-25GEA 0 GBP
COBBBB-2786-25G COBBBB-2786-5G COBBBB-2786-1G
2,4-Dimethylphenylboronic acid
2,4-Dimethylphenylboronic acid
Formula: C₈H₁₁BO₂
MW: 149.98 g/mol
MDL Number: MFCD02683101
CAS Number: 55499-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR