2,4-Dimethoxyphenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BB-2667-5G
BB-2667-25G
BB-2667-1G
BB-2667-100G
COBBBB-2667-5GEA
0
GBP
COBBBB-2667-5G
COBBBB-2667-25G
COBBBB-2667-1G
COBBBB-2667-100G
2,4-Dimethoxyphenylboronic acid
2,4-Dimethoxyphenylboronic acid
Formula:
C₈H₁₁BO₄ MW: 181.98 g/mol |
MDL Number:
MFCD01074590 CAS Number: 133730-34-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...