2,4-Dibromothiazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,4-dibromo-1,3-thiazole

HC-6657-1G HC-6657-5G HC-6657-25G
COBBHC-6657-1GEA 0 GBP
COBBHC-6657-1G COBBHC-6657-5G COBBHC-6657-25G
2,4-Dibromothiazole
2,4-Dibromothiazole
Formula: C₃HBr₂NS
MDL Number: MFCD01318994
CAS Number: 4175-77-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR