2,4-Dibromopyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OS-7460-5G OS-7460-25G OS-7460-1G
COBBOS-7460-5GEA 0 GBP
COBBOS-7460-5G COBBOS-7460-25G COBBOS-7460-1G
2,4-Dibromopyridine
2,4-Dibromopyridine
Formula: C₅H₃Br₂N
MW: 236.89 g/mol
MDL Number: MFCD01859720
CAS Number: 58530-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR