2,4,6-Trimethoxybenzaldehyde

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0036-5G OT-0036-25G OT-0036-100G
COBBOT-0036-5GEA 0 GBP
COBBOT-0036-5G COBBOT-0036-25G COBBOT-0036-100G
2,4,6-Trimethoxybenzaldehyde
2,4,6-Trimethoxybenzaldehyde
Formula: C₁₀H₁₂O₄
MW: 196.2 g/mol
Boiling Pt: 291 °C (1013 hPa)
Melting Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00003313
CAS Number: 830-79-5
EINECS: 212-598-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR