2,4,6-Trichlorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-2362-1G BB-2362-5G BB-2362-25G
COBBBB-2362-1GEA 0 GBP
COBBBB-2362-1G COBBBB-2362-5G COBBBB-2362-25G
2,4,6-Trichlorophenylboronic acid
2,4,6-Trichlorophenylboronic acid
Formula: C₆H₄BCl₃O₂
MW: 225.26 g/mol
MDL Number: MFCD01074621
CAS Number: 73852-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR