Pinacol
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OS-7485-100G
OS-7485-25G
OS-7485-500G
COBBOS-7485-100GEA
0
GBP
COBBOS-7485-100G
COBBOS-7485-25G
COBBOS-7485-500G
Pinacol
Pinacol
Formula:
C₆H₁₄O₂ Boiling Pt: 174 °C (1013 hPa) Melting Pt: 40…41 °C Density: 0.967 g/cm³ (20 °C) Flash Pt: 77 °C |
MDL Number:
MFCD00004462 CAS Number: 76-09-5 EINECS: 200-933-5 Merck Index: 13,07521 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...