2,3-Difluorobenzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OS-7591-5G OS-7591-100G OS-7591-25G
COBBOS-7591-5GEA 0 GBP
COBBOS-7591-5G COBBOS-7591-100G COBBOS-7591-25G
2,3-Difluorobenzoic acid
2,3-Difluorobenzoic acid
Formula: C₇H₄F₂O₂
MW: 158.1 g/mol
MDL Number: MFCD00010267
CAS Number: 4519-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR