2,3-Dichloropyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PY-7069-100G PY-7069-25G PY-7069-5G
COBBPY-7069-100GEA 0 GBP
COBBPY-7069-100G COBBPY-7069-25G COBBPY-7069-5G
2,3-Dichloropyridine
2,3-Dichloropyridine
Formula: C₅H₃Cl₂N
MW: 147.99 g/mol
Boiling Pt: 202.5…203.5 °C (1013 hPa)
Melting Pt: 70…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00006229
CAS Number: 2402-77-9
EINECS: 219-281-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR