2,3-Dichloro-5-(trifluoromethyl)pyridine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CFF610.1
004746-100G
004746-25G
CHMPCFF610.1EA
0
GBP
CHMPCFF610.1
FLUO004746-100G
FLUO004746-25G
2,3-Dichloro-5-(trifluoromethyl)pyridine
2,3-Dichloro-5-(trifluoromethyl)pyridine
Formula:
C₆H₂Cl₂F₃N Boiling Pt: 176 °C (1013 hPa) |
MDL Number:
MFCD00042243 CAS Number: 69045-84-7 EINECS: 410-340-5 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...