2,2,3,3,4,4,5,5-Octafluoro-1-pentanol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1H,1H,5H-Octafluoro-1-pentanol

006918-100G 006918-500G 006918-1KG
FLUO006918-100GEA 0 GBP
FLUO006918-100G FLUO006918-500G FLUO006918-1KG
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol
Formula: C₅H₄F₈O
Boiling Pt: 141 °C (1013 hPa)
Melting Pt: < –50 °C
Density: 1,663 g/cm³ (20 °C)
MDL Number: MFCD00039631
CAS Number: 355-80-6
EINECS: 206-593-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR