2-(Trifluoromethyl)thiobenzamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

021350-1G 021350-5G
FLUO021350-1GEA 0 GBP
FLUO021350-1G FLUO021350-5G
2-(Trifluoromethyl)thiobenzamide
2-(Trifluoromethyl)thiobenzamide
Formula: C₈H₆F₃NS
MDL Number: MFCD04973326
CAS Number: 886496-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR