2-(Trifluoromethyl)pyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OS-7572-100G OS-7572-5G OS-7572-25G
COBBOS-7572-100GEA 0 GBP
COBBOS-7572-100G COBBOS-7572-5G COBBOS-7572-25G
2-(Trifluoromethyl)pyridine
2-(Trifluoromethyl)pyridine
Formula: C₆H₄F₃N
Boiling Pt: 138…139 °C (1013 hPa)
MDL Number: MFCD02183562
CAS Number: 368-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR