2-(Trifluoromethoxy)benzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OS-7502-100G OS-7502-25G OS-7502-5G
COBBOS-7502-100GEA 0 GBP
COBBOS-7502-100G COBBOS-7502-25G COBBOS-7502-5G
2-(Trifluoromethoxy)benzoic acid
2-(Trifluoromethoxy)benzoic acid
Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
MDL Number: MFCD00052325
CAS Number: 1979-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR