2-(Diphenylphosphino)benzoic acid ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

B22389.03 B22389.06 B22389.14
B22389.03EA 51.5 GBP
B22389.03 B22389.06 B22389.14
2-(Diphenylphosphino)benzoic acid ≥97%
2-(Diphenylphosphino)benzoic acid

Purity: 97% CAS No.: 17261-28-8 Molecular Formula: C19H15O2P Formula Weight: 306.31 Melting Point: 174-181° MDL No.: MFCD00674024

Formula: C₁₉H₁₅O₂P
Melting Pt: 174…181 °C
Storage Temperature: Ambient
MDL Number: MFCD00674024
CAS Number: 17261-28-8
EINECS: 241-293-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR