1-Methyl-1H-pyrazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 1-Methylpyrazole

HI-1675-5G HI-1675-100G HI-1675-25G
COBBHI-1675-5GEA 0 GBP
COBBHI-1675-5G COBBHI-1675-100G COBBHI-1675-25G
1-Methyl-1H-pyrazole
1-Methyl-1H-pyrazole
Formula: C₄H₆N₂
MW: 82.11 g/mol
Boiling Pt: 127 °C (1013 hPa)
Density: 0,993 g/cm³ (20 °C)
MDL Number: MFCD00144943
CAS Number: 930-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR