1-Methylpiperazine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OS-7588-25G
OS-7588-100G
COBBOS-7588-25GEA
0
GBP
COBBOS-7588-25G
COBBOS-7588-100G
1-Methylpiperazine
1-Methylpiperazine
Formula:
C₅H₁₂N₂ MW: 100.16 g/mol Boiling Pt: 138 °C (1013 hPa) Melting Pt: –6 °C Density: 0.9085 g/cm³ (20 °C) Flash Pt: 34 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005966 CAS Number: 109-01-3 EINECS: 203-639-5 UN: 2734 ADR: 8,II |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...