3-Methoxy-N-methylaniline hydrochloride
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2924-5G
HC-2924-25G
HC-2924-1G
COBBHC-2924-5GEA
0
GBP
COBBHC-2924-5G
COBBHC-2924-25G
COBBHC-2924-1G
3-Methoxy-N-methylaniline hydrochloride
3-Methoxy-N-methylaniline hydrochloride
Formula:
C₈H₁₂ClNO |
MDL Number:
MFCD13196469 CAS Number: 26926-55-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...