1-Isoquinolinecarboxylic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Isoquinoline-1-carboxylic acid
CA-4490-25G
CA-4490-5G
CA-4490-100G
COBBCA-4490-25GEA
0
GBP
COBBCA-4490-25G
COBBCA-4490-5G
COBBCA-4490-100G
1-Isoquinolinecarboxylic acid
1-Isoquinolinecarboxylic acid
Formula:
C₁₀H₇NO₂ Boiling Pt: 345 °C (1013 hPa) Melting Pt: 161 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006901 CAS Number: 486-73-7 EINECS: 207-639-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...