2-Nitro-4-(trifluoromethyl)phenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009266-1G 009266-5G
FLUO009266-1GEA 0 GBP
FLUO009266-1G FLUO009266-5G
2-Nitro-4-(trifluoromethyl)phenol
2-Nitro-4-(trifluoromethyl)phenol
Formula: C₇H₄F₃NO₃
MDL Number: MFCD00009791
CAS Number: 400-99-7
EINECS: 206-927-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR