α,α,α-Trifluoro-p-cresol
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
OS-7653-25G
OS-7653-5G
OS-7653-100G
COBBOS-7653-25GEA
0
GBP
COBBOS-7653-25G
COBBOS-7653-5G
COBBOS-7653-100G
α,α,α-Trifluoro-p-cresol
α,α,α-Trifluoro-p-cresol
Formula:
C₇H₅F₃O Boiling Pt: 185 °C (1013 hPa) Melting Pt: 45…47 °C Density: 1,3 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002363 CAS Number: 402-45-9 EINECS: 206-945-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...