4-Nitro-3-(trifluoromethyl)phenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006617-1G 006617-5G 006617-25G
FLUO006617-1GEA 0 GBP
FLUO006617-1G FLUO006617-5G FLUO006617-25G
4-Nitro-3-(trifluoromethyl)phenol
4-Nitro-3-(trifluoromethyl)phenol
Formula: C₇H₄F₃NO₃
Boiling Pt: 135…138 °C (0.01 mmHg)
Melting Pt: 81 °C
MDL Number: MFCD00014714
CAS Number: 88-30-2
EINECS: 201-818-2
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR