2,4,6-Tris(dimethylaminomethyl)phenol

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Tris-2,6-dimethylaminomethylphenol, 2,6-Tris-N,N-dimethylaminomethylfenol, DMP-30, Epon® DMP-30 substitute

36975.01 36975.03
SERA36975.01EA 16.53 GBP
SERA36975.01 SERA36975.03
2,4,6-Tris(dimethylaminomethyl)phenol
2,4,6-Tris(dimethylaminomethyl)phenol
Formula: C₁₅H₂₇N₃O
MW: 265.4 g/mol
Boiling Pt: 130…135 °C (1 mmHg)
Density: 0,974 g/cm³ (15 °C)
MDL Number: MFCD00008330
CAS Number: 90-72-2
EINECS: 202-013-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR