2-(Trifluoromethoxy)phenol

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(Trifluorormethoxy)phenol, α,α,α-Trifluoro-2-hydroxyanisole

OS-7613-5G OS-7613-25G OS-7613-100G
COBBOS-7613-5GEA 0 GBP
COBBOS-7613-5G COBBOS-7613-25G COBBOS-7613-100G
2-(Trifluoromethoxy)phenol
2-(Trifluoromethoxy)phenol
Formula: C₇H₅F₃O₂
MDL Number: MFCD00236324
CAS Number: 32858-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR