p,α,α,α-Tetrafluoroanisole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-(Trifluoromethoxy)-1-fluorobenzene, 1-Fluoro-4-(trifluoromethoxy)benzene, α,α,α,4-Tetrafluoroanisole, 4-(Trifluoromethoxy)fluorobenzene
OT-0520-5G
OT-0520-100G
OT-0520-25G
COBBOT-0520-5GEA
0
GBP
COBBOT-0520-5G
COBBOT-0520-100G
COBBOT-0520-25G
p,α,α,α-Tetrafluoroanisole
p,α,α,α-Tetrafluoroanisole
Formula:
C₇H₄F₄O Boiling Pt: 104…105 °C (1013 hPa) Density: 1,323 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00040835 CAS Number: 352-67-0 EINECS: 206-523-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...