3-Fluoro-4-nitrophenol
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
AN-1020-5G
AN-1020-25G
AN-1020-1G
COBBAN-1020-5GEA
0
GBP
COBBAN-1020-5G
COBBAN-1020-25G
COBBAN-1020-1G
3-Fluoro-4-nitrophenol
3-Fluoro-4-nitrophenol
Formula:
C₆H₄FNO₃ MW: 157.1 g/mol Melting Pt: 90…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041251 CAS Number: 394-41-2 EINECS: 206-895-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...