2-Fluoro-5-(trifluoromethyl)phenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006431-5G 006431-25G
FLUO006431-5GEA 0 GBP
FLUO006431-5G FLUO006431-25G
2-Fluoro-5-(trifluoromethyl)phenol
2-Fluoro-5-(trifluoromethyl)phenol
Formula: C₇H₄F₄O
Boiling Pt: 146 °C (1013 hPa)
Density: 1.436 g/cm³ (20 °C)
MDL Number: MFCD00061286
CAS Number: 141483-15-0
UN: 2920
ADR: 8,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR