2-Fluoro-6-methoxyphenol

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0874-1G OT-0874-5G
COBBOT-0874-1GEA 0 GBP
COBBOT-0874-1G COBBOT-0874-5G
2-Fluoro-6-methoxyphenol
2-Fluoro-6-methoxyphenol
Formula: C₇H₇FO₂
MW: 142.13 g/mol
MDL Number: MFCD00075140
CAS Number: 73943-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR