3'-Fluoro-4'-methoxyacetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(3-Fluoro-4-methoxyphenyl)-1-ethanone

006261-25G 006261-5G 006261-100G
FLUO006261-25GEA 0 GBP
FLUO006261-25G FLUO006261-5G FLUO006261-100G
3'-Fluoro-4'-methoxyacetophenone
3'-Fluoro-4'-methoxyacetophenone
Formula: C₉H₉FO₂
MW: 168.17 g/mol
Boiling Pt: 147…148 °C (20 mmHg)
Melting Pt: 92 °C
MDL Number: MFCD00026219
CAS Number: 455-91-4
EINECS: 207-253-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR