2'-Hydroxy-5'-methyl-3'-nitroacetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

012079-25G 012079-5G
FLUO012079-25GEA 0 GBP
FLUO012079-25G FLUO012079-5G
2'-Hydroxy-5'-methyl-3'-nitroacetophenone
2'-Hydroxy-5'-methyl-3'-nitroacetophenone
Formula: C₉H₉NO₄
MW: 195.17 g/mol
MDL Number: MFCD00192216
CAS Number: 66108-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR