4-Fluoro-2-(trifluoromethyl)benzonitrile
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Cyano-5-fluorobenzotrifluoride
OS-7487-25G
OS-7487-100G
COBBOS-7487-25GEA
0
GBP
COBBOS-7487-25G
COBBOS-7487-100G
4-Fluoro-2-(trifluoromethyl)benzonitrile
4-Fluoro-2-(trifluoromethyl)benzonitrile
Formula:
C₈H₃F₄N MW: 189.11 g/mol Boiling Pt: 52…55 °C (15 mmHg) Melting Pt: 43…45 °C Density: 1.35 g/cm³ (20 °C) |
MDL Number:
MFCD00061283 CAS Number: 194853-86-6 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...