3-Fluoro-4-methoxybenzonitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4103-25G CA-4103-100G CA-4103-5G
COBBCA-4103-25GEA 0 GBP
COBBCA-4103-25G COBBCA-4103-100G COBBCA-4103-5G
3-Fluoro-4-methoxybenzonitrile
3-Fluoro-4-methoxybenzonitrile
Formula: C₈H₆FNO
MDL Number: MFCD00215835
CAS Number: 331-62-4
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR