2,3-Difluoro-4-methoxybenzonitrile
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4350-5G
CA-4350-1G
COBBCA-4350-5GEA
0
GBP
COBBCA-4350-5G
COBBCA-4350-1G
2,3-Difluoro-4-methoxybenzonitrile
2,3-Difluoro-4-methoxybenzonitrile
Formula:
C₈H₅F₂NO |
MDL Number:
MFCD03428596 CAS Number: 256417-12-6 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...