5-Chloro-2-nitrobenzotrifluoride
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AN-1147-5G
AN-1147-25G
COBBAN-1147-5GEA
0
GBP
COBBAN-1147-5G
COBBAN-1147-25G
5-Chloro-2-nitrobenzotrifluoride
5-Chloro-2-nitrobenzotrifluoride
Formula:
C₇H₃ClF₃NO₂ Boiling Pt: 222 °C (1013 hPa) Melting Pt: 21…22 °C Density: 1.526 g/cm³ (20 °C) |
MDL Number:
MFCD00007298 CAS Number: 118-83-2 EINECS: 204-280-7 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...