1-Chloro-4-iodobenzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2465-100G HC-2465-25G
COBBHC-2465-100GEA 0 GBP
COBBHC-2465-100G COBBHC-2465-25G
1-Chloro-4-iodobenzene
1-Chloro-4-iodobenzene
Formula: C₆H₄ClI
Boiling Pt: 226 °C (1013 hPa)
Melting Pt: 57 °C
Density: 1,886 g/cm³ (57 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001053
CAS Number: 637-87-6
EINECS: 211-305-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR