4-Chloro-3-(trifluoromethyl)benzonitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Chloro-5-cyanobenzotrifluoride

CA-4408-5G CA-4408-25G CA-4408-100G
COBBCA-4408-5GEA 0 GBP
COBBCA-4408-5G COBBCA-4408-25G COBBCA-4408-100G
4-Chloro-3-(trifluoromethyl)benzonitrile
4-Chloro-3-(trifluoromethyl)benzonitrile
Formula: C₈H₃ClF₃N
MW: 205.57 g/mol
Boiling Pt: 120…125 °C (20 mmHg)
Melting Pt: 66…68 °C
MDL Number: MFCD00190117
CAS Number: 1735-54-2
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR