3-Chlorobiphenyl (PCB No. 2)

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-1122-5G OT-1122-1G
COBBOT-1122-5GEA 0 GBP
COBBOT-1122-5G COBBOT-1122-1G
3-Chlorobiphenyl (PCB No. 2)
3-Chlorobiphenyl (PCB No. 2)
Formula: C₁₂H₉Cl
MW: 188.66 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 16 °C
Density: 1.1579 g/cm³ (25 °C)
MDL Number: MFCD00013633
CAS Number: 2051-61-8
EINECS: 218-126-1
UN: 2315
ADR: 9,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR