5-Chloro-2-nitrophenol

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-1202-5G OT-1202-25G OT-1202-1G
COBBOT-1202-5GEA 0 GBP
COBBOT-1202-5G COBBOT-1202-25G COBBOT-1202-1G
5-Chloro-2-nitrophenol
5-Chloro-2-nitrophenol
Formula: C₆H₄ClNO₃
MW: 173.56 g/mol
MDL Number: MFCD09260849
CAS Number: 611-07-4
EINECS: 210-249-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR