2-Chloro-6-fluoro-3-methoxyphenol

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0639-5G OT-0639-1G
COBBOT-0639-5GEA 0 GBP
COBBOT-0639-5G COBBOT-0639-1G
2-Chloro-6-fluoro-3-methoxyphenol
2-Chloro-6-fluoro-3-methoxyphenol
Formula: C₇H₆ClFO₂
MW: 176.57 g/mol
MDL Number: MFCD09832266
CAS Number: 1017777-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR