3-Chloro-4-hydroxybenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

017248-25G 017248-5G
FLUO017248-25GEA 0 GBP
FLUO017248-25G FLUO017248-5G
3-Chloro-4-hydroxybenzonitrile
3-Chloro-4-hydroxybenzonitrile
Formula: C₇H₄ClNO
MW: 153.57 g/mol
MDL Number: MFCD01567246
CAS Number: 2315-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR