2-Chloro-4-nitroanisole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Chloro-2-methoxy-5-nitrobenzene

AN-1300-5G AN-1300-25G AN-1300-100G
COBBAN-1300-5GEA 0 GBP
COBBAN-1300-5G COBBAN-1300-25G COBBAN-1300-100G
2-Chloro-4-nitroanisole
2-Chloro-4-nitroanisole
Formula: C₇H₆ClNO₃
MW: 187.58 g/mol
MDL Number: MFCD00060681
CAS Number: 4920-79-0
EINECS: 225-547-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR