5-Chloro-2-(trifluoromethyl)phenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

004473-1G 004473-5G 004473-25G
FLUO004473-1GEA 0 GBP
FLUO004473-1G FLUO004473-5G FLUO004473-25G
5-Chloro-2-(trifluoromethyl)phenol
5-Chloro-2-(trifluoromethyl)phenol
Formula: C₇H₄ClF₃O
MDL Number: MFCD00042217
CAS Number: 35852-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR