1-Chloro-2,6-difluoro-3-nitrobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Chloro-2,4-difluoronitrobenzene, 2-Chloro-1,3-difluoro-4-nitrobenzene

013797-25G 013797-5G
FLUO013797-25GEA 0 GBP
FLUO013797-25G FLUO013797-5G
1-Chloro-2,6-difluoro-3-nitrobenzene
1-Chloro-2,6-difluoro-3-nitrobenzene
Formula: C₆H₂ClF₂NO₂
MW: 193.54 g/mol
MDL Number: MFCD00042201
CAS Number: 3847-58-3
EINECS: 411-980-8
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR