4-Bromo-2-nitrophenol
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AN-1280-25G
AN-1280-5G
AN-1280-100G
COBBAN-1280-25GEA
0
GBP
COBBAN-1280-25G
COBBAN-1280-5G
COBBAN-1280-100G
4-Bromo-2-nitrophenol
4-Bromo-2-nitrophenol
Formula:
C₆H₄BrNO₃ MW: 218.01 g/mol Boiling Pt: 259 °C (1013 hPa) Melting Pt: 90…94 °C |
MDL Number:
MFCD00082540 CAS Number: 7693-52-9 EINECS: 231-707-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...