1-Bromo-4-fluoro-3-nitrobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001657-25G 001657-100G
FLUO001657-25GEA 0 GBP
FLUO001657-25G FLUO001657-100G
1-Bromo-4-fluoro-3-nitrobenzene
1-Bromo-4-fluoro-3-nitrobenzene
Formula: C₆H₃BrFNO₂
Boiling Pt: 145…147 °C (35 mmHg)
Melting Pt: 19 °C
Density: 1.786 g/cm³ (20 °C)
MDL Number: MFCD00129165
CAS Number: 364-73-8
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR