2-Bromo-α,α,α,5-tetrafluorotoluene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OS-7716-25G OS-7716-100G
COBBOS-7716-25GEA 0 GBP
COBBOS-7716-25G COBBOS-7716-100G
2-Bromo-α,α,α,5-tetrafluorotoluene
2-Bromo-α,α,α,5-tetrafluorotoluene
Formula: C₇H₃BrF₄
MDL Number: MFCD00040937
CAS Number: 40161-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR